accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOFFFFOOH3C

CAS RN 377-71-9

IUPAC Name: 1,4-diethyl 2,2,3,3-tetrafluorobutanedioate
Synonyms: 1,4-diethyl 2,2,3,3-tetrafluorobutanedioate
Molecular Weight (g/mol): 246.16
Molecular Formula: C8H10F4O4
InChi Key: BLZSSBPZHBFNBN-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)C(F)(F)C(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results