accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OONH3C

CAS RN 37619-24-2

IUPAC Name: methyl 1-methyl-1H-pyrrole-2-carboxylate
Synonyms: methyl 1-methylpyrrole-2-carboxylate
Molecular Weight (g/mol): 139.15
Molecular Formula: C7H9NO2
InChi Key: APHVGKYWHWFAQV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CN1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results