accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CMgBr

CAS RN 3761-92-0

IUPAC Name: magnesium(2+) hexan-1-ide bromide
Synonyms: magnesium(2+) hexan-1-ide bromide
Molecular Weight (g/mol): 189.38
Molecular Formula: C6H13BrMg
InChi Key: LZFCBBSYZJPPIV-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CCCCC[CH2-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results