accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFOOOFFFF

CAS RN 376-68-1

IUPAC Name: hexafluorooxane-2,6-dione
Synonyms: hexafluoroglutaric anhydride
Molecular Weight (g/mol): 222.04
Molecular Formula: C5F6O3
InChi Key: IHYAGCYJVNHXCT-UHFFFAOYSA-N
SMILES: FC1(F)C(=O)OC(=O)C(F)(F)C1(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results