accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 37535-58-3

H3CH3CCH3OONH(R)NOO

CAS RN 37535-58-3

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(pyridin-4-yl)propanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-(pyridin-4-yl)propanoate
Molecular Weight (g/mol): 265.29
Molecular Formula: C13H17N2O4
InChi Key: FNYWDMKESUACOU-SNVBAGLBSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=NC=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)