accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 374790-97-3

HOBOHBrF

CAS RN 374790-97-3

IUPAC Name: (4-bromo-3-fluorophenyl)boronic acid
Synonyms: 4-bromo-3-fluorophenylboronic acid
Molecular Weight (g/mol): 218.82
Molecular Formula: C6H5BBrFO2
InChi Key: ACLQPRPXJMWADE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Br)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results