accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 37451-68-6

NCH3H3CH3CCH3Cl

CAS RN 37451-68-6

IUPAC Name: tetrabutylazanium hydrate chloride
Synonyms: tetrabutylammonium ion hydrate chloride
Molecular Weight (g/mol): 295.94
Molecular Formula: C16H38ClNO
InChi Key: FODWRUPJCKASBN-UHFFFAOYSA-M
SMILES: O.[Cl-].CCCC[N+](CCCC)(CCCC)CCCC
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)