accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3728-20-9

IUPAC Name: hydrogen methyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate chloride
Synonyms: hydrogen methyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate chloride
Molecular Weight (g/mol): 231.68
Molecular Formula: C10H14ClNO3
InChi Key: VXYFARNRGZWHTJ-SBSPUUFOSA-N
SMILES: [H+].[Cl-].COC(=O)[C@H](N)CC1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results