accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 37245-77-5

NH2NFeFeFeFeFeFe

CAS RN 37245-77-5

IUPAC Name: ferrioazanide pentairon azanide
Synonyms: ferrioazanide pentairon azanide
Molecular Weight (g/mol): 365.1
Molecular Formula: Fe6H2N2
InChi Key: ILMFDGFXPJCFQW-UHFFFAOYSA-N
SMILES: [NH2-].[Fe].[Fe].[Fe].[Fe].[Fe].[N-]=[Fe]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results