CAS RN 37174-63-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
S-Methyl N,N-Diethylthiocarbamate, TRC
CAS: 37174-63-3 Molecular Formula: C6 H13 N O S Molecular Weight (g/mol): 147.24 Synonym: Disulfiram Impurity 3 IUPAC Name: S-methyl N,N-diethylcarbamothioate SMILES: CCN(CC)C(=O)SC