accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFBrFFFFBr

CAS RN 371-83-5

IUPAC Name: 1,5-dibromo-1,1,3,3,5,5-hexafluoropentane
Synonyms: 1,5-dibromo-1,1,3,3,5,5-hexafluoropentane
Molecular Weight (g/mol): 337.89
Molecular Formula: C5H4Br2F6
InChi Key: XYMBGGTVDWPIBA-UHFFFAOYSA-N
SMILES: FC(F)(Br)CC(F)(F)CC(F)(F)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results