accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3704-28-7

IUPAC Name: methyl 2-(methylsulfanyl)benzoate
Synonyms: methyl 2-(methylsulfanyl)benzoate
Molecular Weight (g/mol): 182.24
Molecular Formula: C9H10O2S
InChi Key: CPQDZXPLQXZJGF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1SC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results