accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 37026-88-3

BrH3CH3NH3CH3C

CAS RN 37026-88-3

IUPAC Name: 5-butylnonan-5-aminium bromide
Synonyms: 5-butylnonan-5-aminium bromide
Molecular Weight (g/mol): 280.29
Molecular Formula: C13H30BrN
InChi Key: FQUHTZCWXDFLBQ-UHFFFAOYSA-N
SMILES: [Br-].CCCCC([NH3+])(CCCC)CCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results