accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOPOOH3COH3C

CAS RN 3699-67-0

IUPAC Name: ethyl 3-(diethoxyphosphoryl)propanoate
Synonyms: ethyl 3-(diethoxyphosphoryl)propanoate
Molecular Weight (g/mol): 238.22
Molecular Formula: C9H19O5P
InChi Key: JMJWCUOIOKBVNQ-UHFFFAOYSA-N
SMILES: CCOC(=O)CCP(=O)(OCC)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results