accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 369403-08-7

IUPAC Name: 1-{[(tert-butoxy)carbonyl]amino}-4-hydroxycyclohexane-1-carboxylate
Synonyms: 1-[(tert-butoxycarbonyl)amino]-4-hydroxycyclohexane-1-carboxylate
Molecular Weight (g/mol): 258.3
Molecular Formula: C12H20NO5
InChi Key: DRBVVUBYEBFRKS-UHFFFAOYSA-M
SMILES: CC(C)(C)OC(=O)NC1(CCC(O)CC1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results