accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONOFF

CAS RN 369-34-6

IUPAC Name: 1,2-difluoro-4-nitrobenzene
Synonyms: 1,2-difluoro-4-nitrobenzene
Molecular Weight (g/mol): 159.09
Molecular Formula: C6H3F2NO2
InChi Key: RUBQQRMAWLSCCJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C(F)=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)