accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 368870-05-7

IUPAC Name: 5-(2-methyl-1,3-thiazol-4-yl)-1,2-oxazole-3-carboxylic acid
Synonyms: 5-(2-methyl-1,3-thiazol-4-yl)-1,2-oxazole-3-carboxylic acid
Molecular Weight (g/mol): 210.21
Molecular Formula: C8H6N2O3S
InChi Key: QAXQRHWAJNDTCV-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC(=NO1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results