accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONSOOH

CAS RN 368869-97-0

IUPAC Name: 2-(2,3-dihydro-1-benzofuran-5-yl)-1,3-thiazole-4-carboxylic acid
Synonyms: 2-(2,3-dihydro-1-benzofuran-5-yl)-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 247.27
Molecular Formula: C12H9NO3S
InChi Key: NOUOFPSRFBTJCH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC(=N1)C1=CC2=C(OCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results