accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OHNOOO(E)

CAS RN 36847-94-6

IUPAC Name: (2E)-3-[(2-methoxyphenyl)carbamoyl]prop-2-enoate
Synonyms: (2E)-3-[(2-methoxyphenyl)carbamoyl]prop-2-enoate
Molecular Weight (g/mol): 220.21
Molecular Formula: C11H10NO4
InChi Key: HGLMERMZXFKZOL-VOTSOKGWSA-M
SMILES: COC1=CC=CC=C1NC(=O)\C=C\C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results