accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 36847-11-7

BrBrNBrN

CAS RN 36847-11-7

IUPAC Name: 2,4,6-tribromopyrimidine
Synonyms: 2,4,6-tribromopyrimidine
Molecular Weight (g/mol): 316.78
Molecular Formula: C4HBr3N2
InChi Key: AHEYFWKLKMOHCI-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=NC(Br)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)