accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COFOOH

CAS RN 367-83-9

IUPAC Name: 2-fluoro-5-methoxybenzoic acid
Synonyms: 2-fluoro-5-methoxybenzoic acid
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: REDSLTKMNCCQBC-UHFFFAOYSA-N
SMILES: COC1=CC(C(O)=O)=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results