accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

RhBFFFF

CAS RN 36620-11-8

IUPAC Name: bis(bicyclo[2.2.1]hepta-2,5-diene) tetrafluoroboranuide rhodium
Synonyms: bis(norbornadiene) rhodium tetrafluoroborate
Molecular Weight (g/mol): 373.99
Molecular Formula: C14H16BF4Rh
InChi Key: ZWPWNBFOFVFMPP-UHFFFAOYSA-N
SMILES: [Rh].F[B-](F)(F)F.C1C2C=CC1C=C2.C1C2C=CC1C=C2
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)