accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 366-63-2

FONH

CAS RN 366-63-2

IUPAC Name: 4-fluoro-N-phenylbenzamide
Synonyms: 4-fluoro-N-phenylbenzamide
Molecular Weight (g/mol): 215.23
Molecular Formula: C13H10FNO
InChi Key: FIEGRQABVBXYQT-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results