accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOSNNOOH3CH3CCH3

CAS RN 365413-31-6

IUPAC Name: tert-butyl 4-[4-(ethoxycarbonyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate
Synonyms: tert-butyl 4-[4-(ethoxycarbonyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate
Molecular Weight (g/mol): 340.44
Molecular Formula: C16H24N2O4S
InChi Key: LBRUPXMVELEXIV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CSC(=N1)C1CCN(CC1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results