accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNHSH2NHCl

CAS RN 36518-76-0

IUPAC Name: 2-imino-2,5-dihydro-1,3-thiazol-4-amine hydrochloride
Synonyms: 2-imino-5H-1,3-thiazol-4-amine hydrochloride
Molecular Weight (g/mol): 151.61
Molecular Formula: C3H6ClN3S
InChi Key: ANDLBJXZBLTCDP-UHFFFAOYSA-N
SMILES: Cl.NC1=NC(=N)SC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results