accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 36404-30-5

H3COClCl

CAS RN 36404-30-5

IUPAC Name: 1,2-dichloro-4-methoxybenzene
Synonyms: 1,2-dichloro-4-methoxybenzene
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: VISJRVXHPNMYRH-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)