accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNNOCl

CAS RN 36401-55-5

IUPAC Name: 5-methyl-2-phenyl-2H-1,2,3-triazole-4-carbonyl chloride
Synonyms: 5-methyl-2-phenyl-1,2,3-triazole-4-carbonyl chloride
Molecular Weight (g/mol): 221.64
Molecular Formula: C10H8ClN3O
InChi Key: UJYBUZMRRLFXGM-UHFFFAOYSA-N
SMILES: CC1=NN(N=C1C(Cl)=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results