accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2BrBr

CAS RN 3638-73-1

IUPAC Name: 2,5-dibromoaniline
Synonyms: 2,5-dibromoaniline
Molecular Weight (g/mol): 250.92
Molecular Formula: C6H5Br2N
InChi Key: WRTAZRGRFBCKBU-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CC=C1Br
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)