accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COFNH2F

CAS RN 363-47-3

IUPAC Name: 3,5-difluoro-4-methoxyaniline
Synonyms: 3,5-difluoro-4-methoxyaniline
Molecular Weight (g/mol): 159.14
Molecular Formula: C7H7F2NO
InChi Key: POVSDXPEJZMSEJ-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(N)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results