accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 362003-83-6

OHNNSNHO

CAS RN 362003-83-6

IUPAC Name: N-(2-benzamido-1,3-benzothiazol-6-yl)adamantane-1-carboxamide
Synonyms: N-(2-benzamido-1,3-benzothiazol-6-yl)adamantane-1-carboxamide
Molecular Weight (g/mol): 431.55
Molecular Formula: C25H25N3O2S
InChi Key: MVSSJPGNLQPWSW-UHFFFAOYSA-N
SMILES: O=C(NC1=NC2=CC=C(NC(=O)C34CC5CC(CC(C5)C3)C4)C=C2S1)C1=CC=CC=C1
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results