accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NHNBr

CAS RN 3610-42-2

IUPAC Name: 2-(5-bromo-1H-indol-3-yl)ethan-1-amine
Synonyms: 2-(5-bromo-1H-indol-3-yl)ethanamine
Molecular Weight (g/mol): 239.12
Molecular Formula: C10H11BrN2
InChi Key: CGHUQJRRADEHTQ-UHFFFAOYSA-N
SMILES: NCCC1=CNC2=CC=C(Br)C=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results