accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 36051-68-0

NH2NON(R)O(R)OPOOOPOOOPHOHOO(S)HO(R)OHNaNa

CAS RN 36051-68-0

IUPAC Name: disodium 4-amino-1-[5-({[(hydrogen phosphonatooxy)(hydroxy)phosphoryl phosphonato]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms: disodium 4-amino-1-[5-({[hydrogen phosphonatooxy(hydroxy)phosphoryl phosphonato]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]pyrimidin-2-one
Molecular Weight (g/mol): 527.12
Molecular Formula: C9H14N3Na2O14P3
InChi Key: NFQMDTRPCFJJND-UHFFFAOYNA-L
SMILES: [Na+].[Na+].NC1=NC(=O)N(C=C1)C1OC(COP([O-])(=O)OP(O)(=O)OP(O)([O-])=O)C(O)C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)