accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3CH3H3COONHHO

CAS RN 36016-38-3

IUPAC Name: tert-butyl N-hydroxycarbamate
Synonyms: tert-butyl N-hydroxycarbamate
Molecular Weight (g/mol): 133.15
Molecular Formula: C5H11NO3
InChi Key: DRDVJQOGFWAVLH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NO
Quantity 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results