accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNCH3H3CH3COOFFF

CAS RN 35895-70-6

IUPAC Name: tetrabutylazanium trifluoroacetate
Synonyms: tetrabutylammonium ion trifluoroacetate
Molecular Weight (g/mol): 355.49
Molecular Formula: C18H36F3NO2
InChi Key: WTEXQPWIUJQYJQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)