accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 356531-63-0

CH3OHNF

CAS RN 356531-63-0

IUPAC Name: N-[(4-fluorophenyl)methyl]-3-methoxyaniline
Synonyms: N-[(4-fluorophenyl)methyl]-3-methoxyaniline
Molecular Weight (g/mol): 231.27
Molecular Formula: C14H14FNO
InChi Key: LFWQQAPNOAPZNW-UHFFFAOYSA-N
SMILES: COC1=CC=CC(NCC2=CC=C(F)C=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results