accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHFFFFFFFFHO

CAS RN 355-74-8

IUPAC Name: 2,2,3,3,4,4,5,5-octafluorohexane-1,6-diol
Synonyms: 2,2,3,3,4,4,5,5-octafluorohexane-1,6-diol
Molecular Weight (g/mol): 262.1
Molecular Formula: C6H6F8O2
InChi Key: NHEKBXPLFJSSBZ-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)CO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)