accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFFClFClFFF

CAS RN 355-20-4

IUPAC Name: 2,3-dichloro-1,1,1,2,3,4,4,4-octafluorobutane
Synonyms: 2,3-dichloro-1,1,1,2,3,4,4,4-octafluorobutane
Molecular Weight (g/mol): 270.93
Molecular Formula: C4Cl2F8
InChi Key: LXANZHXWGZWFAC-UHFFFAOYNA-N
SMILES: FC(F)(F)C(F)(Cl)C(F)(Cl)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results