accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 35461-99-5

OHOO

CAS RN 35461-99-5

IUPAC Name: 3-(furan-2-yl)benzoic acid
Synonyms: 3-(furan-2-yl)benzoic acid
Molecular Weight (g/mol): 188.18
Molecular Formula: C11H8O3
InChi Key: RQVVFGRDMHDHNI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results