accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2N

CAS RN 3544-25-0

IUPAC Name: 2-(4-aminophenyl)acetonitrile
Synonyms: (4-amino-phenyl)-acetonitrile
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: YCWRFIYBUQBHJI-UHFFFAOYSA-N
SMILES: NC1=CC=C(CC#N)C=C1
Boiling Point 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)