accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 35376-44-4

OOH

CAS RN 35376-44-4

IUPAC Name: 3-hydroxy-5-phenylcyclohex-2-en-1-one
Synonyms: 3-hydroxy-5-phenylcyclohex-2-en-1-one
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: ZFVDSCASCRHXCP-UHFFFAOYNA-N
SMILES: OC1=CC(=O)CC(C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results