accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 35344-11-7

CH3CCCCCH3CH3CCH3CH3CH3CCCCCH3CH3CCH3CH3COCOCOCOFeFe

CAS RN 35344-11-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 494.19
Molecular Formula: C24H30Fe2O4
InChi Key: UXGBSDPRTITVSY-UHFFFAOYSA-N
SMILES: [Fe].[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].C[C]1[C](C)[C](C)[C](C)[C]1C.C[C]1[C](C)[C](C)[C](C)[C]1C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results