accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3529-10-0

H3CNCH3NH2

CAS RN 3529-10-0

IUPAC Name: (4-aminobutyl)dimethylamine
Synonyms: (4-aminobutyl)dimethylamine
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: GCOWZPRIMFGIDQ-UHFFFAOYSA-N
SMILES: CN(C)CCCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results