accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BSOCH3HOOH

CAS RN 352530-25-7

IUPAC Name: (5-formyl-4-methylthiophen-2-yl)boronic acid
Synonyms: 5-formyl-4-methylthiophen-2-ylboronic acid
Molecular Weight (g/mol): 169.99
Molecular Formula: C6H7BO3S
InChi Key: QEIXJCOHLUKRPO-UHFFFAOYSA-N
SMILES: CC1=C(SC(=C1)B(O)O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results