accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 352530-24-6

IUPAC Name: [4-(ethanesulfonyl)phenyl]boronic acid
Synonyms: 4-(ethanesulfonyl)phenylboronic acid
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H11BO4S
InChi Key: CVBNDDGEVPNUNA-UHFFFAOYSA-N
SMILES: CCS(=O)(=O)C1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results