accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HClNH2BOHHO

CAS RN 352525-94-1

IUPAC Name: hydrogen [3-(aminomethyl)phenyl]boronic acid chloride
Synonyms: hydrogen 3-(aminomethyl)phenylboronic acid chloride
Molecular Weight (g/mol): 187.43
Molecular Formula: C7H11BClNO2
InChi Key: NPTBTFRGCBFYPZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results