accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OPOOOH

CAS RN 35193-64-7

IUPAC Name: 13-hydroxy-12,14-dioxa-13λ⁵-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(15),2,4,6,8,10,16,18,20,22-decaen-13-one
Synonyms: 13-hydroxy-12,14-dioxa-13λ⁵-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(15),2,4,6,8,10,16,18,20,22-decaen-13-one
Molecular Weight (g/mol): 348.29
Molecular Formula: C20H13O4P
InChi Key: JEHUZVBIUCAMRZ-UHFFFAOYSA-N
SMILES: OP1(=O)OC2=CC=C3C=CC=CC3=C2C2=C(O1)C=CC1=CC=CC=C21
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results