accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NOOH

CAS RN 35166-33-7

IUPAC Name: (5-methyl-1,2-oxazol-3-yl)methanol
Synonyms: (5-methyl-1,2-oxazol-3-yl)methanol
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: MDYHWQQHEWDJKR-UHFFFAOYSA-N
SMILES: CC1=CC(CO)=NO1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)