accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClCH3OSiNH3CH3CCH3OH3COH3C

CAS RN 35141-36-7

IUPAC Name: trimethyl[3-(trimethoxysilyl)propyl]azanium chloride
Synonyms: trimethyl[3-(trimethoxysilyl)propyl]azanium chloride
Molecular Weight (g/mol): 257.83
Molecular Formula: C9H24ClNO3Si
InChi Key: FYZFRYWTMMVDLR-UHFFFAOYSA-M
SMILES: [Cl-].CO[Si](CCC[N+](C)(C)C)(OC)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results