accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOBOHFN

CAS RN 351019-18-6

IUPAC Name: (6-fluoropyridin-3-yl)boronic acid
Synonyms: 6-fluoropyridin-3-ylboronic acid
Molecular Weight (g/mol): 140.91
Molecular Formula: C5H5BFNO2
InChi Key: OJBYZWHAPXIJID-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(F)N=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)