accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 351003-46-8

IUPAC Name: 3-bromo-5-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 3-bromo-5-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 323.51
Molecular Formula: C7H3BrClF3O2S
InChi Key: YBUJCZFWJSUTSN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(Br)=C1)S(Cl)(=O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results